Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL
Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL
Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL
Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL
Thermo Scientific Chemicals
Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL
CAS: 7789-02-8 | CrN3O9 | 238.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF088059.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialChromium
Name NoteAAS Standard Solution
CAS7789-02-8
Health Hazard 1H314-H332-H335
Health Hazard 2When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).