Cadmium, AAS standard solution, Specpure™ Cd 1000μg/mL
Cadmium, AAS standard solution, Specpure™ Cd 1000μg/mL
Thermo Scientific Chemicals
Cadmium, AAS standard solution, Specpure™ Cd 1000μg/mL
Cd in 5% HNO3
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF088056.AP
Price (MYR)
510.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
510.00
EA
Add to cart
Specifications
Chemical Name or MaterialCadmium, 1000μg/mL
Name NoteCd 1000μg/mL
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H350-H361-H314
May cause cancer.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.