Boron, AAS solution standard, Specpure™ B 1000μg/mL
Boron, AAS solution standard, Specpure™ B 1000μg/mL
Thermo Scientific Chemicals
Boron, AAS solution standard, Specpure™ B 1000μg/mL
H3BO3 in H2O
Have Questions?
Catalog NumberQuantity
ALF088055.AE100 mL
Catalog number ALF088055.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialBoron, AAS standard solution
Name NoteB 1000μg/mL
Health Hazard 1H360FD
Health Hazard 3P201-P202-P281-P308+P313-P501c