Tellurium(IV) oxide, 99.99% (metals basis)
Tellurium(IV) oxide, 99.99% (metals basis)
Tellurium(IV) oxide, 99.99% (metals basis)
Tellurium(IV) oxide, 99.99% (metals basis)
Thermo Scientific Chemicals
Tellurium(IV) oxide, 99.99% (metals basis)
CAS: 7446-07-3 | O2Te | 159.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF087813.22
Price (MYR)
2,422.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
2,422.00
EA
Add to cart
Specifications
Chemical Name or MaterialTellurium(IV) oxide
Melting Point733°C
CAS7446-07-3
Health Hazard 1H317-H332-H360
Health Hazard 3P201-P202-P261-P271-P272-P280g-P281-P302+P352-P304+P340-P308+P313-P333+P313-P363-P501c
View more
Tellurium(IV) oxide is also used in devices which can convert light into sound (acousto-optic material). Glasses made of tellurium oxide have high refractive indices and transmit into mid-IR region. It is used to make glasses with special properties. It is useful in optical waveguides and optical fiber amplification.