Ferrocene, 99%
Ferrocene, 99%
Ferrocene, 99%
Thermo Scientific Chemicals
Ferrocene, 99%
CAS: 102-54-5 | C10H10Fe | 186.04 g/mol
Have Questions?
Catalog NumberQuantity
ALF087202.1850 g
Catalog number ALF087202.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialFerrocene
CAS102-54-5
Health Hazard 1H228-H302
Health Hazard 2GHS H Statement
H228-H302
Flammable solid.
Harmful if swallowed.
Health Hazard 3P210-P240-P241-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
View more
Ferrocene is used as a catalyst for carbon nanotubes and for rocket propellants. It is also used in the fuel for petrol engines, gasoline antiknock, luminous sensitive materials and exhibit anticancer activity. It acts as a photosensitive catalyst and promote the photochemical degadation of plastic films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only