Dimethyl diselenide, 99%
Dimethyl diselenide, 99%
Dimethyl diselenide, 99%
Dimethyl diselenide, 99%
Thermo Scientific Chemicals
Dimethyl diselenide, 99%
CAS: 7101-31-7 | C2H6Se2 | 188.012 g/mol
Have Questions?
Catalog NumberQuantity
ALF068119.031 g
Catalog number ALF068119.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDimethyl diselenide
CAS7101-31-7
Health Hazard 1H226
Health Hazard 2GHS H Statement
H301-H331-H226-H373
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
View more
Dimethyl diselenide is a powerful reagent used for identification of distonic radical cations