Antimony, AAS standard solution, Specpure™ Sb 1000μg/mL
Antimony, AAS standard solution, Specpure™ Sb 1000μg/mL
Antimony, AAS standard solution, Specpure™ Sb 1000μg/mL
Antimony, AAS standard solution, Specpure™ Sb 1000μg/mL
Thermo Scientific Chemicals
Antimony, AAS standard solution, Specpure™ Sb 1000μg/mL
Sb in 30% HCl
Have Questions?
Catalog NumberQuantity
ALF047197.AE100 mL
Catalog number ALF047197.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialAntimony
Name NoteAAS standard solution, Specpure™
CAS7732-18-5
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314-H335