Iron(II,III) oxide, nanopowder, 97%
Iron(II,III) oxide, nanopowder, 97%
Iron(II,III) oxide, nanopowder, 97%
Iron(II,III) oxide, nanopowder, 97%
Thermo Scientific Chemicals
Iron(II,III) oxide, nanopowder, 97%
CAS: 1317-61-9 | Fe3O4 | 231.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALF047141.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIron(II,III) oxide
Melting Point1538°C
CAS1317-61-9
ColorBlack
Health Hazard 1Warning
View more
Iron(II,III) oxide is mainly used as a black-pigment-catalyst in the Haber process and in the water gas shift reaction. It is used as a contrast-agent in MRI scanning and as a pigment in magnetic applications, polishing compounds, cosmetics, medicines, polymers, rubber, fillers, building, construction, appliances and magnetic inks.