Diaion™ HP20, synthetic adsorbent resin, highly porous type
Diaion™ HP20, synthetic adsorbent resin, highly porous type
Thermo Scientific Chemicals
Diaion™ HP20, synthetic adsorbent resin, highly porous type
Have Questions?
Catalog NumberQuantity
ALF046488.A11 kg
Catalog number ALF046488.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialDiaion™ HP20, synthetic adsorbent resin
Physical FormBeads
Name NoteHighly porous type
Recommended StorageAmbient temperatures
TSCANo
View more
Diaion™ HP20 is used as a filler in chromatography columns due to its extremely large pores. It is considered as an effective adsorbent to eliminate toxic effects in fermentation and separation of traces heavy metal ions. It is also useful for the purification of peptides, proteins, fermentation products, polyphenols and cephalosporin C. It helps to remove bitter tasting substances from fruit juices.