Platinum Combustion Boat, Top length 200mm, width 16mm, Ht 18mm, Base Thickness 0.20mm, Capacity = 52.5mL, Thermo Scientific Chemicals
Platinum Combustion Boat, Top length 200mm, width 16mm, Ht  18mm, Base Thickness 0.20mm, Capacity = 52.5mL, Thermo Scientific Chemicals
Actual product may vary
Platinum Combustion Boat, Top length 200mm, width 16mm, Ht 18mm, Base Thickness 0.20mm, Capacity = 52.5mL, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF046473.KT1 Ea.
Catalog number ALF046473.KT
Price (MYR)
-
Quantity:
1 Ea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity1 Ea.
Unit SizeEA