Zeolite Y, ammonium
Zeolite Y, ammonium
Zeolite Y, ammonium
Thermo Scientific Chemicals
Zeolite Y, ammonium
CAS: 1318-02-1 | Al2O5Si | 162.043 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALF045867.36
Price (MYR)
2,378.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
2,378.00
EA
Add to cart
Specifications
Chemical Name or MaterialZeolite Y, ammonium
CAS1318-02-1
ColorWhite
Recommended StorageAmbient temperatures
Physical FormPowder
View more
Zeolite Y is a useful metal aluminum silicate for proteomics research. It is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products. It is also used as a molecular sieve and as an adsorbent of a wide variety of cations, such as Na+, K+, Ca2+, Mg2+ and others. Zeolite is used in the development of a variety of bioreactor devices and in the determination of serum triglycerides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products
RUO – Research Use Only