Silicon(IV) nitride, amorphous, nanopowder, 98.5+%
Silicon(IV) nitride, amorphous, nanopowder, 98.5+%
Silicon(IV) nitride, amorphous, nanopowder, 98.5+%
Silicon(IV) nitride, amorphous, nanopowder, 98.5+%
Thermo Scientific Chemicals
Silicon(IV) nitride, amorphous, nanopowder, 98.5+%
CAS: 12033-89-5 | N4Si3 | 140.28 g/mol
Have Questions?
Catalog NumberQuantity
ALF045611.065 g
Catalog number ALF045611.06
Price (MYR)
390.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
390.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilicon (IV) nitride
Melting Point1900°C (sublimation)
Name NoteAmorphous
CAS12033-89-5
ColorGray