Polyethyleneimine, branched, M.W. 1,200, 99%
Polyethyleneimine, branched, M.W. 1,200, 99%
Polyethyleneimine, branched, M.W. 1,200, 99%
Polyethyleneimine, branched, M.W. 1,200, 99%
Thermo Scientific Chemicals
Polyethyleneimine, branched, M.W. 1,200, 99%
CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol
Have Questions?
Catalog NumberQuantity
ALF045594.22100 g
Catalog number ALF045594.22
Price (MYR)
1,014.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,014.00
EA
Add to cart
Specifications
Chemical Name or MaterialPolyethyleneimine
ColorYellow
Name NoteBranched, M.W. 1,200
CAS9002-98-6
Physical FormLiquid
View more
Polyethyleneimine acts as a protein precipitant used to purify proteins. It is used as a chelating agent and as a scavenger for aldehydes and oxides. It is also used in detergents, paper industry, dyes, printing inks and in water treatment.