Amberlite™ IRN-78, ion exchange resin, nuclear grade
Amberlite™ IRN-78, ion exchange resin, nuclear grade
Amberlite™ IRN-78, ion exchange resin, nuclear grade
Amberlite™ IRN-78, ion exchange resin, nuclear grade
Thermo Scientific Chemicals
Amberlite™ IRN-78, ion exchange resin, nuclear grade
Strongly basic anion exchange resin | CAS: 11128-95-3 | Styrene-DVB
Have Questions?
Catalog NumberQuantity
ALF045591.36500 g
Catalog number ALF045591.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmberlite™ IRN-78
Physical FormBeads
CAS11128-95-3
Health Hazard 1H318
Health Hazard 3P280-P305+P351+P338-P310
View more
Amberlite™ IRN-78 is widely employed in removing radioactive anions for example 131 Iodine and 133 Iodine from waste systems. It also removes the traces of chloride contamination from reactor coolant systems and controls the boron level in the primary system. At higher pH conditions, it is used to remove anionic impurities from secondary streams in the presence of ammonia, morpholine or other amines.