Polydimethylsiloxane (silicone) emulsion
Polydimethylsiloxane (silicone) emulsion
Thermo Scientific Chemicals
Polydimethylsiloxane (silicone) emulsion
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Catalog number ALF045088.A4
Price (MYR)
1,206.00
EA
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (MYR)
1,206.00
EA
Add to cart
Specifications
Chemical Name or MaterialPolydimethylsiloxane (silicone) emulsion
Physical FormLiquid
Recommended StorageAmbient temperatures
TSCAYes
MDL NumberMFCD00132673
View more
Polydimethylsiloxane (silicone) emulsion is the most widely used silicon-based organic polymer and is particularly known for its unusual rheological (or flow) properties. Further, it is used in contact lenses, medical devices to elastomers and in shampoos (as dimethicone makes hair shiny and slippery). It finds application as an antifoaming agent in food, caulking, lubricants, kinetic sand and heat-resistant tiles. In addition to this, it serves as a critical ingredient in water-repelling coatings such as Rain-X.