Aluminum flake, APS 11 micron, 99.7% (metals basis)
Aluminum flake, APS 11 micron, 99.7% (metals basis)
Aluminum flake, APS 11 micron, 99.7% (metals basis)
Aluminum flake, APS 11 micron, 99.7% (metals basis)
Thermo Scientific Chemicals
Aluminum flake, APS 11 micron, 99.7% (metals basis)
CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Catalog number ALF045031.A7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum
ColorSilver to Gray
CAS7429-90-5
Health Hazard 1Contact with water liberates highly flammable gases. Spontaneously flammable in air.
Health Hazard 2Keep container tightly closed and dry.