Silver Conductive Ink
Silver Conductive Ink
Silver Conductive Ink
Have Questions?
Catalog NumberCapacity (Metric)
ALF044958.065 g
Catalog number ALF044958.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
5 g
Request bulk or custom format
Specifications
Product TypeConductive Ink
Capacity (Metric)5 g
ColorSilver
For Use With (Application)Membrane switch production
Unit SizeEA
This is a conducting screen printable thick film ink for membrane switch production.