Hydroxypropyl methylcellulose
Hydroxypropyl methylcellulose
Hydroxypropyl methylcellulose
Hydroxypropyl methylcellulose
Thermo Scientific Chemicals
Hydroxypropyl methylcellulose
CAS: 9004-65-3 | C56H108O30 | 1261.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Catalog number ALF044779.A1
Price (MYR)
2,329.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
2,329.00
EA
Add to cart
Specifications
Chemical Name or MaterialHydroxypropyl methylcellulose
CAS9004-65-3
Recommended StorageAmbient temperatures
TSCAYes
Molecular FormulaC56H108O30
View more
Hydroxypropyl methylcellulose is used as an ophthalmic lubricant, an emulsifier and a thickening and suspending agent. It is widely used as an excipient in pharmaceutical formulations. It acts as a food additive. Its eye drops are known as artificial tears, which are used to relieve eye dryness and soreness. It finds applications in various fields as tile adhesives, gypsum products, cosmetics, cement renders, contact lenses, detergents and cleaners.