Europium(III) oxide, nanopowder, 99.9% (REO)
Europium(III) oxide, nanopowder, 99.9% (REO)
Europium(III) oxide, nanopowder, 99.9% (REO)
Europium(III) oxide, nanopowder, 99.9% (REO)
Thermo Scientific Chemicals
Europium(III) oxide, nanopowder, 99.9% (REO)
CAS: 1308-96-9 | Eu2O3 | 351.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALF044667.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialEuropium (III) oxide
Melting Point2291°C
CAS1308-96-9
Recommended StorageAmbient temperatures
Physical FormPowder
View more
Europium oxide nanopowder acts as phosphors for lamps. It is used as a fluorescent label in immunoassay for atrazine. It is also used in color TV, X-ray, glass, crystal and other luminescent materials.