Hydrogen chloride, nominally 2.5M in ethanol
Hydrogen chloride, nominally 2.5M in ethanol
Hydrogen chloride, nominally 2.5M in ethanol
Hydrogen chloride, nominally 2.5M in ethanol
Thermo Scientific Chemicals
Hydrogen chloride, nominally 2.5M in ethanol
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Catalog number ALF044605.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrochloric acid
CAS7647-01-0
Health Hazard 1H225-H290-H314-H332-H335
Health Hazard 2GHS H Statement
H225-H331-H314
Highly flammable liquid and vapor.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Health Hazard 3P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
View more
Hydrogen chloride, 2.5M. in ethanol is used in the manufacturing of pharmaceuticals to get a hydrochloride salt. For example, ranitidine base is converted to ranitidine hydrochloride, which is used as anti ulcerative drug.