3-Hydroxy-5-methylisoxazole, 97%
3-Hydroxy-5-methylisoxazole, 97%
3-Hydroxy-5-methylisoxazole, 97%
3-Hydroxy-5-methylisoxazole, 97%
Thermo Scientific Chemicals
3-Hydroxy-5-methylisoxazole, 97%
CAS: 10004-44-1 | C4H5NO2 | 99.089 g/mol
Have Questions?
Catalog NumberQuantity
ALF044450.065 g
Catalog number ALF044450.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Hydroxy-5-methylisoxazole
CAS10004-44-1
Health Hazard 1H302-H318
Health Hazard 2GHS H Statement
H318
Causes serious eye damage.
Health Hazard 3P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
View more
3-Hydroxy-5-methylisoxazole is used in plant growth stimulation activity and seedlings growing. And it is also used in DNA/RNA synthesis inhibitor.