Oxalic acid, anhydrous, 98%
Oxalic acid, anhydrous, 98%
Oxalic acid, anhydrous, 98%
Oxalic acid, anhydrous, 98%
Thermo Scientific Chemicals
Oxalic acid, anhydrous, 98%
CAS: 144-62-7 | C2H2O4 | 90.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALF044410.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialOxalic acid
Name NoteAnhydrous
pH1.3
CAS144-62-7
Melting Point∼185°C (decomposition)
View more
Oxalic acid is used as reducing agent for photographic films, as mordant in dyeing process, as bleaching agent for removal of rust through complex formation, and as grinding agent during polishing of marble. It is used in waste water treatment to remove calcium. The conjugate base of oxalic acid acts as a chelating agent for metal ions. It is used as a catalyst for esterifications in synthetic organic chemistry and as an analytical reagent.