Bicinchoninic acid disodium salt
Bicinchoninic acid disodium salt
Bicinchoninic acid disodium salt
Bicinchoninic acid disodium salt
Thermo Scientific Chemicals
Bicinchoninic acid disodium salt
For determination of Cu and protein assays | CAS: 979-88-4 | C20H10N2Na2O4 | 388.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number ALF044247.09
Price (MYR)
2,104.00
EA
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
2,104.00
EA
Add to cart
Specifications
Chemical Name or MaterialBicinchoninic acid disodium salt
CAS979-88-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
For determination of Cu and protein assays.Bicinchoninic acid disodium salt is useful for the determination of copper. It is also used for the determination of protein concentration.