Praseodymium powder, -40 mesh, 99.5% (REO)
Praseodymium powder, -40 mesh, 99.5% (REO)
Praseodymium powder, -40 mesh, 99.5% (REO)
Praseodymium powder, -40 mesh, 99.5% (REO)
Thermo Scientific Chemicals
Praseodymium powder, -40 mesh, 99.5% (REO)
CAS: 7440-10-0 | Pr | 140.91 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ALF044183.065 gAmpule
Catalog number ALF044183.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Ampule
Request bulk or custom format
Specifications
Chemical Name or MaterialPraseodymium powder
ColorGray to Silver
CAS7440-10-0
Health Hazard 1H228-H500
Health Hazard 2GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only