Gadolinium(III) oxide, nanopowder, 99.99+% (REO)
Gadolinium(III) oxide, nanopowder, 99.99+% (REO)
Gadolinium(III) oxide, nanopowder, 99.99+% (REO)
Gadolinium(III) oxide, nanopowder, 99.99+% (REO)
Thermo Scientific Chemicals
Gadolinium(III) oxide, nanopowder, 99.99+% (REO)
CAS: 12064-62-9 | Gd2O3 | 362.50 g/mol
Have Questions?
Catalog NumberQuantity
ALF044020.1425 g
Catalog number ALF044020.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGadolinium (III) oxide
Melting Point2330°C
CAS12064-62-9
Health Hazard 1H319
Health Hazard 3P264b-P280i-P305+P351+P338
View more
Gadolinium(III) oxide is used as a raw material for various fluorescent compounds, absorption materials in atomic reactions, nuclear fuels, magnetic bubble material and screen-sensitivity increasing material. It is also used in glass and electronic industries. It also acts as a neutron shield, a catalyst, a dielectric ceramic and is used for filament coatings, special glasses, lasers, masers and telecommunication. It also serves as a laboratory reagent. It plays an important role as a potential contrast agent for magnetic resonance imaging (MRI).