Matrix oil, 20 centistoke hydrocarbon oil, Thermo Scientific Chemicals
Matrix oil, 20 centistoke hydrocarbon oil, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Matrix oil, 20 centistoke hydrocarbon oil, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF044012.LV1 gal.
Catalog number ALF044012.LV
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 gal.
Request bulk or custom format
Specifications
Chemical Name or MaterialMatrix oil
Name Note20 centistoke hydrocarbon oil
CAS64742-65-0
Health Hazard 1H350
Health Hazard 2GHS H Statement
H350
May cause cancer.