Aluminum foil, 0.25mm (0.01in) thick, annealed, Puratronic™, 99.9995% (metals basis)
Aluminum foil, 0.25mm (0.01in) thick, annealed, Puratronic™, 99.9995% (metals basis)
Aluminum foil, 0.25mm (0.01in) thick, annealed, Puratronic™, 99.9995% (metals basis)
Aluminum foil, 0.25mm (0.01in) thick, annealed, Puratronic™, 99.9995% (metals basis)
Aluminum foil, 0.25mm (0.01in) thick, annealed, Puratronic™, 99.9995% (metals basis)
Thermo Scientific Chemicals
Aluminum foil, 0.25mm (0.01in) thick, annealed, Puratronic™, 99.9995% (metals basis)
CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Catalog NumberQuantity
ALF043424.FI50 x 50 mm
Catalog number ALF043424.FI
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 x 50 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum foil
Name Note0.25mm (0.01 in.) thick, Annealed, Temper: soft
CAS7429-90-5
Recommended StorageAmbient temperatures
EINECS Number231-072-3