2-Butanol, anhydrous, 99%
2-Butanol, anhydrous, 99%
2-Butanol, anhydrous, 99%
2-Butanol, anhydrous, 99%
Thermo Scientific Chemicals
2-Butanol, anhydrous, 99%
Hygroscopic; H2O <0.01% | CAS: 78-92-2 | C4H10O | 74.12 g/mol
Have Questions?
Catalog NumberQuantity
ALF043315.AK250 mL
Catalog number ALF043315.AK
Price (MYR)
344.00
EA
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
344.00
EA
Add to cart
Specifications
Chemical Name or Material2-Butanol
Melting Point−115°C
Name NoteH2O <0.01%
CAS78-92-2
ColorColorless
View more
2-Butanol is widely used as a solvent for domestic cleaning agents and paint removers. It is also utilized in perfumes and artificial flavors. It plays an important role in the extraction of fish meal to produce fish protein concentrate. It is involved in the production of fruit essences. It plays an essential role in the esterification of acetic acid. It is useful for the derivatization of chlorinated hydroxyfuranones. It is also used as a precursor for methyl ethyl ketone.