Europium(III) acetate hydrate, REacton™, 99.9% (REO)
Europium(III) acetate hydrate, REacton™, 99.9% (REO)
Europium(III) acetate hydrate, REacton™, 99.9% (REO)
Europium(III) acetate hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals
Europium(III) acetate hydrate, REacton™, 99.9% (REO)
CAS: 62667-64-5 | C6H9EuO6 | 329.10 g/mol
Have Questions?
Catalog NumberQuantity
ALF042878.1425 g
Catalog number ALF042878.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialEuropium(III) acetate hydrate
CAS62667-64-5
ColorWhite
Recommended StorageAmbient temperatures
Physical FormPowder
View more
Europium(III) acetate hydrate is used as fluorescent powder and as a chemical reagent.