Divinylbenzene, 55%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol
Divinylbenzene, 55%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol
Divinylbenzene, 55%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol
Divinylbenzene, 55%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol
Thermo Scientific Chemicals
Divinylbenzene, 55%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol
Crosslinking monomer | CAS: 1321-74-0 | C10H10 | 130.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALF042805.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDivinylbenzene
Name NoteMixture of isomers, Stabilized with 1000ppm 4-tert-butylcatechol
CAS1321-74-0
Health Hazard 1H227-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
View more
Used as a co-monomer in the production of ion exchange beads and gel chromatography. As a monomer in the production of synthetic rubber and casting resins. Also used in polymeric core shell nanoparticles and as a crosslinking agent. It may be used as a monomer for the synthesis of highly crosslinked macroporous styrene/DVB polymers. It may also be used to synthesize monodispersed poly-DVB microspheres.