Potassium hydroxide, 0.1N Standardized Solution in methanol
Potassium hydroxide, 0.1N Standardized Solution in methanol
Potassium hydroxide, 0.1N Standardized Solution in methanol
Potassium hydroxide, 0.1N Standardized Solution in methanol
Thermo Scientific Chemicals
Potassium hydroxide, 0.1N Standardized Solution in methanol
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Catalog NumberQuantity
ALF042734.K21 L
Catalog number ALF042734.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium hydroxide Standardized Solution
ColorColorless
CAS1310-58-3
Health Hazard 1H225-H301+H311+H331-H370
Health Hazard 2GHS H Statement
H225-H311-H331-H370-H315-H319
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.