Silica gel, wide pore, 150 angstroms
Silica gel, wide pore, 150 angstroms
Silica gel, wide pore, 150 angstroms
Have Questions?
Catalog NumberQuantity
ALF042726.A75 kg
Catalog number ALF042726.A7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Request bulk or custom format
Specifications
Particle Size-100/200 Mesh
Pore Size150 Å
PackagingDrum
pH6.5 to 7.5
Quantity5 kg
View more
Silica gel, wide pore is mainly used for the dehydrated purification of the industrial gases, the clearance of the organic acids and high polymers in the insulative oil, the purification and separation of the medicines. It is also used in column chromatography without applied pressure (gravity pulled).