Aluminum oxide, alpha-phase, 99.9% (metals basis)
Aluminum oxide, alpha-phase, 99.9% (metals basis)
Aluminum oxide, alpha-phase, 99.9% (metals basis)
Aluminum oxide, alpha-phase, 99.9% (metals basis)
Thermo Scientific Chemicals
Aluminum oxide, alpha-phase, 99.9% (metals basis)
CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Catalog NumberQuantity
ALF042571.22100 g
Catalog number ALF042571.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Boiling Point2980°C
Chemical Name or MaterialAluminum oxide
Melting Point2045°C
CAS1344-28-1
ColorWhite
View more
Alumium oxide is widely used in a variety of plastics, rubber, ceramics, refractory products for reinforcement toughening. It is also useful to produce artificial ruby, sapphire, yttrium aluminum garnet. It is useful in the field of fiber fabric products and high pressure sodium lamp as far-infrared emission and thermal insulation materials. It is useful for the C single crystal detectors. It plays an important role for the hydrogenation of aromatic hydrocarbons.