Silica gel 60, 0.040-0.063mm (230-400 mesh), 99.5% (metals basis)
Silica gel 60, 0.040-0.063mm (230-400 mesh), 99.5% (metals basis)
Silica gel 60, 0.040-0.063mm (230-400 mesh), 99.5% (metals basis)
Silica gel 60, 0.040-0.063mm (230-400 mesh), 99.5% (metals basis)
Thermo Scientific Chemicals
Silica gel 60, 0.040-0.063mm (230-400 mesh), 99.5% (metals basis)
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Catalog number ALF042570.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialSilica gel 60
Physical FormPowder, -230+400 Mesh,S.A. 500-600m
CAS7631-86-9
Health Hazard 1H373: May cause damage to organs through prolonged or repeated exposure.
Recommended StorageAmbient temperatures
View more
Column chromatographySilica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.