Total Organic Carbon (TOC), standard solution, Specpure™ 1000μg/mL
Total Organic Carbon (TOC), standard solution, Specpure™ 1000μg/mL
Thermo Scientific Chemicals
Total Organic Carbon (TOC), standard solution, Specpure™ 1000μg/mL
KHC8H4O4 in H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF042562.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialTotal Organic Carbon
Name Note1000μg/mL
Recommended StorageAmbient temperatures
OdorCharacteristic
TSCAYes
View more
Total Organic Carbon (TOC), is used as a standard solution during the measurement of chemical analysis such as detecting contaminants in drinking water and cooling water. It is also used for analysis of water to be used for semiconductor manufacturing and pharmaceutical use.