Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 2000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 2000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 2000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 2000
Thermo Scientific Chemicals
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 2000
CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol
Have Questions?
Catalog NumberQuantity
ALF042499.K21 L
Catalog number ALF042499.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialPolydimethylsiloxane, trimethylsiloxy terminated
ColorColorless
Name NoteM.W. 2,000
CAS9016-00-6
Physical FormLiquid
View more
Polydimethylsiloxane, trimethylsiloxy terminated is widely used as a common surfactant. It is an active component of defoamers which suppresses the formation of foams and is a critical ingredient in water-repelling coatings as an active silicone fluid. It finds application in optic telecommunications, biomedical research, elasticity of polymer networks, adhesive materials and in skin-moisturizing lotions. Furthermore, it is used as a filler fluid in breast implants and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.