(3-Mercaptopropyl)methyldimethoxysilane, 95%
(3-Mercaptopropyl)methyldimethoxysilane, 95%
(3-Mercaptopropyl)methyldimethoxysilane, 95%
(3-Mercaptopropyl)methyldimethoxysilane, 95%
Thermo Scientific Chemicals
(3-Mercaptopropyl)methyldimethoxysilane, 95%
CAS: 31001-77-1 | C6H16O2SSi | 180.337 g/mol
Have Questions?
Catalog NumberQuantity
ALF042426.AK250 mL
Catalog number ALF042426.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Boiling Point96°C (30mmHg)
Chemical Name or Material(3-Mercaptopropyl)methyldimethoxysilane
CAS31001-77-1
ColorColorless
Health Hazard 1H227-H302-H317
View more
(3-Mercaptopropyl)methyldimethoxysilane is used as coupling agent to improve the adhesion of sulfur cured elastomers (polysulfide, polyurethane sealants) to inorganic fillers (such as silica, glass, clay, mica), fiberglass and surfaces. It is also used as a pretreatment on minerals or added at the time of compounding and to improve low-rolling resistance in silica-reinforced tire tread compounds.