Buffer solution, pH 6.86 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable
Buffer solution, pH 6.86 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable
Buffer solution, pH 6.86 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable
Buffer solution, pH 6.86 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable
Thermo Scientific Chemicals
Buffer solution, pH 6.86 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Catalog NumberQuantity
ALF042419.AP500 mL
Catalog number ALF042419.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS1336-21-6
Specifications
Chemical Name or MaterialBuffer solution
CAS1336-21-6
ColorColorless
Recommended StorageAmbient temperatures
TSCAYes