n-Octane-d{18}, 99% (Isotopic)
n-Octane-d{18}, 99% (Isotopic)
n-Octane-d{18}, 99% (Isotopic)
n-Octane-d{18}, 99% (Isotopic)
Thermo Scientific Chemicals
n-Octane-d{18}, 99% (Isotopic)
CAS: 17252-77-6 | C8H18 | 132.342 g/mol
Have Questions?
Catalog NumberQuantity
ALF042339.031 g
Catalog number ALF042339.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Materialn-Octane-d{18}
Density0.815 g/mL
Flash Point15.6°C (60°F)
Physical FormLiquid
Quantity1 g