Polyethylene oxide, M.W. 300,000
Polyethylene oxide, M.W. 300,000
Polyethylene oxide, M.W. 300,000
Polyethylene oxide, M.W. 300,000
Thermo Scientific Chemicals
Polyethylene oxide, M.W. 300,000
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF042237.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyethylene oxide
ColorWhite
Name NoteM.W. 300,000
pH5 to 7
CAS25322-68-3
View more
Polyethylene oxide, M.W. 300,000 is used to impart viscosity to and modify flow of aqueous solutions.