Nafion∣r D-520 dispersion, 5% w/w in water and 1-propanol, ∣>1.00 meq/g exchange capacity
Nafion∣r D-520 dispersion, 5% w/w in water and 1-propanol, ∣>1.00 meq/g exchange capacity
Nafion∣r D-520 dispersion, 5% w/w in water and 1-propanol, ∣>1.00 meq/g exchange capacity
Nafion∣r D-520 dispersion, 5% w/w in water and 1-propanol, ∣>1.00 meq/g exchange capacity
Nafion∣r D-520 dispersion, 5% w/w in water and 1-propanol, ∣>1.00 meq/g exchange capacity
Preparation of thin films and coatings; proton exchange membranes | CAS: 31175-20-9 | C9HF17O5S | 544.135 g/mol
Have Questions?
Catalog NumberQuantity
ALF042118.AE100 mL
Catalog number ALF042118.AE
Price (MYR)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialNafion™ D-520 dispersion
ColorColorless to Yellow
Name Note≥0.92meq/g exchange capacity
CAS31175-20-9
Physical FormLiquid
View more
Preparation of thin films and coatings; proton exchange membraneNafion™ D-520 dispersion, 5% w/w in water and 1-propanol is used in the fabrication of thin films and coating formulations for fuel cell membranes, in catalyst coatings, sensors, and various electrochemical applications.