Toluene, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles
Toluene, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles
Toluene, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles
Toluene, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals
Toluene, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles
CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
Catalog NumberQuantity
ALF041841.K21 L
Catalog number ALF041841.K2
Price (MYR)
458.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
458.00
EA
Add to cart
Specifications
Chemical Name or MaterialToluene, anhydrous
Melting Point°C to 93°C
CAS108-88-3
Health Hazard 1H225-H304-H315-H319-H336-H361d-H373
Health Hazard 3P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
View more
Toluene is widely used as an organic solvent in various industries like paints, pharmaceuticals, dyes, agrochemicals and polymers. It is used as an additive in gasoline and improves octane ratings, as a fullerene indicator, in paint thinners and silicone sealants, as a coolant in sodium cold traps used in nuclear reactor system loops and as a fuel for both two-stroke and four-stroke engines. It is also used as a raw material in the production of benzene, benzaldehyde, benzoic acid, toluene diisocyanate, benzyl-tert-butyl peroxide, xylene and many other chemical products. It plays an important role in biochemistry experiments to break open red blood cells in order to extract haemoglobin as well as used in the removal of cocaine from coca leaves to make coca-cola syrup. Further, it used as a standard material for chromatography and as an analytical reagent.