Polyvinylpyrrolidone, cross linked
Polyvinylpyrrolidone, cross linked
Polyvinylpyrrolidone, cross linked
Polyvinylpyrrolidone, cross linked
Thermo Scientific Chemicals
Polyvinylpyrrolidone, cross linked
CAS: 25249-54-1 | (C6H9NO)n | 111.14 g/mol
Have Questions?
Catalog NumberQuantity
ALF041631.22100 g
Catalog number ALF041631.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyvinylpyrrolidone
ColorWhite
Name NoteCross linked
CAS25249-54-1
Physical FormPowder
View more
Polyvinylpyrrolidone is used in coatings, printing ink, rubber, glass, leather, cosmetics, soaps, plastics, in paper making and as a stabilizer. It acts as a disintegrant in pharmaceutical tablets and as a fining to extract impurities. It is used to remove polyphenols in beer.