Silver powder, spherical, APS 0.5-1 micron, 99.9% (metals basis)
Silver powder, spherical, APS 0.5-1 micron, 99.9% (metals basis)
Silver powder, spherical, APS 0.5-1 micron, 99.9% (metals basis)
Silver powder, spherical, APS 0.5-1 micron, 99.9% (metals basis)
Thermo Scientific Chemicals
Silver powder, spherical, APS 0.5-1 micron, 99.9% (metals basis)
Form: S.A. 1.5-2.5m2/g | CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF041597.0910 g
Catalog number ALF041597.09
Price (MYR)
549.00
EA
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
549.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilver
ColorSilver
CAS7440-22-4
Health Hazard 1H400-H410: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
AppearanceSilver-colored powder