Cobalt(II) phthalocyanine
Cobalt(II) phthalocyanine
Cobalt(II) phthalocyanine
Cobalt(II) phthalocyanine
Thermo Scientific Chemicals
Cobalt(II) phthalocyanine
CAS: 3317-67-7 | C32H16CoN8 | 571.469 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALF041496.14
Price (MYR)
2,318.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
2,318.00
EA
Add to cart
Specifications
Chemical Name or MaterialCobalt(II) phthalocyanine
CAS3317-67-7
Health Hazard 1H334-H341-H350
Health Hazard 2GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P261-P281-P285-P304+P341-P308+P313-P342+P311-P501c
View more
Cobalt(II) phthalocyanine may be used to develop carbon black based electrocatalysts. CoPC may be used as an end capping agent of a hyperbranched poly(aryl ether ketone) to be used for oxidative decomposition of 2,4,6,-trichlorophenol. CoPC modified carbon paste may be used as indicators for electrocatalytic amperometric measurements. ZnO impregnated with CoPC may be used as a sensitizer to determine the photodegradation of cyanide in aqueous suspension. CoPC catalyzed aerobic regenerations of aldehydes and ketones have been investigated.