Methanol, anhydrous, 99.9%
Methanol, anhydrous, 99.9%
Methanol, anhydrous, 99.9%
Methanol, anhydrous, 99.9%
Thermo Scientific Chemicals
Methanol, anhydrous, 99.9%
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Catalog number ALF041467.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialMethanol
Melting Point°C to 98°C
CAS67-56-1
Health Hazard 1H225-H301+H311+H331-H370-H372
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
View more
Methanol is widely used as a solvent in synthetic organic chemistry. It is also used as a solvent in analytical chemistry, especially for HPLC, LC-MS, GC-MS, ICP-MS and spectrophotometry. It is used in the production of biodiesel, formaldehyde and acetic acid. It is also used as fuel, antifreeze, denaturant for ethanol and an energy carrier for fuel cell application.