Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight
Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight
Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight
Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight
Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight
Thermo Scientific Chemicals
Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight
CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Catalog number ALF041243.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyvinyl alcohol
Name NoteHigh molecular weight, Hydrolyzed
CAS9002-89-5
Physical FormSolid
Recommended StorageAmbient temperatures
View more
Polyvinyl alcohol is used in food industries as a binding and coating agent. Its viscosity favors the application as coating agent to tablets and capsules. Mixed-matrix membrane prepared by dispersion of NaA zeolite particles is used to study the pervaporative separation of water-butanol mixtures. Polyvinyl alcohol /gelatin based biocompatible polymeric scaffolds have been developed for 3D cancer models.