Polyvinyl alcohol, 98-99% hydrolyzed, medium molecular weight
Polyvinyl alcohol, 98-99% hydrolyzed, medium molecular weight
Polyvinyl alcohol, 98-99% hydrolyzed, medium molecular weight
Polyvinyl alcohol, 98-99% hydrolyzed, medium molecular weight
Thermo Scientific Chemicals
Polyvinyl alcohol, 98-99% hydrolyzed, medium molecular weight
CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Catalog number ALF041242.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyvinyl alcohol
Name NoteMedium molecular weight, Hydrolyzed
CAS9002-89-5
Recommended StorageAmbient temperatures
TSCAYes
View more
Polyvinyl alcohol is used in papermaking, textiles, and in a variety of coatings. It is used as an emulsion polymerization aid to make polyvinyl acetate, as a protective colloid, as a film used in the water transfer printing process, as a surfactant for the formation of polymer encapsulated nanobeads, as an embolization agent in medical procedures and as a fixative for specimen collection. It is an important raw material for polyvinyl nitrate (PVN). It is used in protective chemical-resistant gloves, in eye drops (such as artificial tears to treat dry eyes) and in hard contact lens solution as a lubricant and in 3D printing as a support structure that can then be dissolved away.