Amberlite™ IRN-150, ion exchange resin
Amberlite™ IRN-150, ion exchange resin
Thermo Scientific Chemicals
Amberlite™ IRN-150, ion exchange resin
Styrene-DVB; Mix of strongly acidic and basic gel type resin | Styrene-DVB
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
500 g
2 kg
Catalog number ALF040992.A9
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialAmberlite IRN-150
Physical FormBeads
Health Hazard 1H318
Health Hazard 3P280-P305+P351+P338-P310
Recommended StorageAmbient temperatures
View more
Amberlite IRN-150 is used in water treatment and purification in primary side chemical volume control systems (CVS) in PWR nuclear power generating facilities. It is also used in reactor water clean-up and radio active waste systems. It is recommended in any non-regenerable mixed bed application and can also be used for a variety of electroplating industry waste regeneration applications.