Samarium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
Samarium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
Samarium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
Samarium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
Thermo Scientific Chemicals
Samarium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
CAS: 64438-53-5 | C15H21O6Sm | 447.69 g/mol
Have Questions?
Catalog NumberQuantity
ALF040665.1425 g
Catalog number ALF040665.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSamarium(III) 2,4-pentanedionate hydrate
CAS64438-53-5
Recommended StorageAmbient temperatures
Physical FormPowder
Quantity25 g